乙酸钯(II)

供应商: LOCAL DEALS

Synonyms: Pd(OAc)2

R02412980131 R02412980131CD R02412980121
CHLCR02412980131EA 0 CNY
CHLCR02412980131 CHLCR02412980131CD CHLCR02412980121
乙酸钯(II)
乙酸钯(II)
分子式: C₄H₆O₄Pd
分子量: 224.51 g/mol
溶点: 205
MDL编号: MFCD00012453
CAS号: 3375-31-3
EINECS: 222-164-4

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR