乙酸钯(II)

供应商: LOCAL DEALS

R02412910133 R02412910144
CHLCR02412910133EA 0 CNY
CHLCR02412910133 CHLCR02412910144
乙酸钯(II)
乙酸钯(II)
分子式: C₄H₆O₄Pd
溶点: 205 °C
MDL编号: MFCD00012453
CAS号: 3375-31-3
EINECS: 222-164-4

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR