乙腈

供应商: LOCAL DEALS
Danger

Synonyms: 甲基氰, ACN

R00282720024CD R00282720024
CHLCR00282720024CDEA 0 CNY
CHLCR00282720024CD CHLCR00282720024
乙腈
乙腈
分子式: H₃CCN
分子量: 41.05 g/mol
沸点: 81.6
溶点: –45.7 °C
密度: 0.782 g/cm³ (20 °C)
闪点: 2 °C (closed cup)
存储条件: Ambient
MDL编号: MFCD00001878
CAS号: 75-05-8
EINECS: 200-835-2
UN号: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR