Heptane (mixture of isomers)

供应商: LOCAL DEALS
Danger

R01943920016 R01943920124
CHLCR01943920016EA 0 CNY
CHLCR01943920016 CHLCR01943920124
Heptane (mixture of isomers)
Heptane (mixture of isomers)
分子式: H₃C(CH₂)₅CH₃
分子量: 100.2 g/mol
沸点: 97…98 °C (1013 hPa)
溶点: –90.5 °C
密度: 0.68 g/cm³ (20 °C)
闪点: –4 °C
存储条件: Ambient
MDL编号: MFCD00009544
CAS号: 142-82-5
EINECS: 205-563-8
UN号: 1206
ADR: 3 II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dabde8793eb27ef796098b842bf18b9ced8539b8&vid=d30a8083280d1915a43f51d93c49f9b76eabd579&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR