Iodine

供应商: LOCAL DEALS
Danger

R00274020123 R00274020123CD
CHLCR00274020123EA 0 CNY
CHLCR00274020123 CHLCR00274020123CD
Iodine
分子式: I₂
分子量: 253.81 g/mol
沸点: 184.3
溶点: 114
密度: 4.9 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00011355
CAS号: 7553-56-2
EINECS: 231-442-4
UN号: 3495
ADR: 8 III
Merck Index: 13,05036

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR