吡啶-3-硼酸

供应商: LOCAL DEALS

Synonyms: 3-吡啶硼酸

R00828810131 R00828810134
CHLCR00828810131EA 0 CNY
CHLCR00828810131 CHLCR00828810134
吡啶-3-硼酸
吡啶-3-硼酸
分子式: C₅H₆BNO₂
分子量: 122.92 g/mol
MDL编号: MFCD00674177
CAS号: 1692-25-7

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR