4-Chloro-2-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
供应商: TCI
|
Warning
|
Synonyms:
4-Chloro-2-(trifluoromethyl)benzeneboronic acid, (4-Chloro-2-(trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-chloro-2-(trifluoromethyl)phenyl)borane
C2601-1G
C2601-5G
TCIAC2601-1GEA
0
CNY
TCIAC2601-1G
TCIAC2601-5G
4-Chloro-2-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-2-(trifluoromethyl)phenylboronic acid
分子式:
C₇H₅BClF₃O₂ 分子量: 224.37 |
MDL编号:
MFCD04973086 CAS号: 313545-41-4 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 109.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...