(R)-(-)-1-苯基-1,2-乙二醇 ≥98.0% (通过GC)
供应商: TCI
P1150-1G P1150-5G
TCIAP1150-1GEA 0 CNY
TCIAP1150-1G TCIAP1150-5G
(R)-(-)-1-苯基-1,2-乙二醇 ≥98.0% (通过GC)
(R)-(-)-1-苯基-1,2-乙二醇
分子式: C₈H₁₀O₂
沸点: 82°C
溶点: 67…69 °C
MDL编号: MFCD00064262
CAS号: 16355-00-3
Merck Index: 14,08861
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa59820f81d0865da62361666544a000594e5b6e&vid=aa59820f81d0865da62361666544a000594e5b6e&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 69.0 deg-C
Specific rotation [a]20/D -35.0 to -42.0 deg (C=3, H2O)