2,4′-二氯苯乙酮 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-Chloro-1-(4-chlorophenyl)ethanone, α,4-Dichloroacetophenone, 2,4'-Dichloroacetophenone

C1306-25G C1306-5G
TCIAC1306-25GEA 0 CNY
TCIAC1306-25G TCIAC1306-5G
2,4′-二氯苯乙酮 ≥98.0% (by GC)
2,4′-二氯苯乙酮
分子式: C₈H₆Cl₂O
沸点: 270°C
溶点: 98 °C
MDL编号: MFCD00018926
CAS号: 937-20-2
EINECS: 213-323-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fed5a926990f1036daec42e385856dd12a484b06&vid=fed5a926990f1036daec42e385856dd12a484b06&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 100.0 to 103.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR