乙二胺四乙酸钠盐

供应商: LOCAL DEALS

Synonyms: 四乙酸二氨基乙烯, EDTA

R00196120023 R00196120023CD
CHLCR00196120023EA 0 CNY
CHLCR00196120023 CHLCR00196120023CD
乙二胺四乙酸钠盐
乙二胺四乙酸钠盐
分子式: C₁₀H₁₄N₂Na₂O₈
分子量: 336.21 g/mol
存储条件: Ambient
MDL编号: MFCD00070672
CAS号: 139-33-3
EINECS: 205-358-3
Merck Index: 12,03556

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR