Carbon

供应商: LOCAL DEALS

Synonyms: Charcoal

R00629720111 R00629720011
CHLCR00629720111EA 0 CNY
CHLCR00629720111 CHLCR00629720011
Carbon
Carbon
分子式: C
分子量: 12.01 g/mol
沸点: ∼4000 °C (1013 hPa)
溶点: 3727
密度: 3.65 g/cm³ (20 °C)
MDL编号: MFCD00133992
CAS号: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR