二甲胺盐酸盐

供应商: LOCAL DEALS
Warning

R01111530121 R01111530121CD
CHLCR01111530121EA 0 CNY
CHLCR01111530121 CHLCR01111530121CD
二甲胺盐酸盐
二甲胺盐酸盐
分子式: C₂H₈ClN
溶点: 170…173 °C
MDL编号: MFCD00012477
CAS号: 506-59-2
EINECS: 208-046-5
Merck Index: 13,03255

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR