N-碘代琥珀酰亚胺

供应商: LOCAL DEALS
Warning

Synonyms: 1-Iodopyrrolidine-2,5-dione

R00545420121 R00545420124
CHLCR00545420121EA 0 CNY
CHLCR00545420121 CHLCR00545420124
N-碘代琥珀酰亚胺
N-碘代琥珀酰亚胺
分子式: C₄H₄INO₂
分子量: 224.99 g/mol
沸点: 356
溶点: 200…202 °C
密度: 2.245 g/cm³ (20 °C)
存储条件: Refrigerator
MDL编号: MFCD00005512
CAS号: 516-12-1
EINECS: 208-221-6
Merck Index: 13,05065

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR