6-甲氧基-8-硝基喹啉 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
M2057-25G
M2057-5G
TCIAM2057-25GEA
0
CNY
TCIAM2057-25G
TCIAM2057-5G
6-甲氧基-8-硝基喹啉 ≥98.0% (by GC)
6-甲氧基-8-硝基喹啉
分子式:
C₁₀H₈N₂O₃ 分子量: 204.1800 g/mol |
MDL编号:
MFCD00006802 CAS号: 85-81-4 EINECS: 201-633-7 |
|
产品技术规格
Purity (GC) | min. 98.0 % |
Melting point | 158.0 to 162.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...