1,4-Dibromo-2,5-diiodobenzene ≥96.0% (by GC)
供应商: TCI
|
Warning
|
D3904-25G
D3904-5G
TCIAD3904-25GEA
0
CNY
TCIAD3904-25G
TCIAD3904-5G
1,4-Dibromo-2,5-diiodobenzene ≥96.0% (by GC)
1,4-Dibromo-2,5-diiodobenzene
分子式:
C₆H₂Br₂I₂ 分子量: 487.6970 g/mol |
MDL编号:
MFCD00082898 CAS号: 63262-06-6 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 165.0 to 169.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...