4'-Methoxy-2,2,2-trifluoroacetophenone ≥98.0% (通过GC)

供应商: TCI
Warning

T1986-25G T1986-5G
TCIAT1986-25GEA 0 CNY
TCIAT1986-25G TCIAT1986-5G
4'-Methoxy-2,2,2-trifluoroacetophenone ≥98.0% (通过GC)
4'-Methoxy-2,2,2-trifluoroacetophenone
分子式: C₉H₇F₃O₂
MDL编号: MFCD00018061
CAS号: 711-38-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14e0497018d81dfc0edb31de0a1785f112479bd7&vid=14e0497018d81dfc0edb31de0a1785f112479bd7&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR