4'-Fluoro-3'-(trifluoromethyl)acetophenone ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: 1-(4-Fluoro-3-(trifluoromethyl)phenyl)ethanone

F0808-1G F0808-5G
TCIAF0808-1GEA 0 CNY
TCIAF0808-1G TCIAF0808-5G
4'-Fluoro-3'-(trifluoromethyl)acetophenone ≥97.0% (by GC)
4'-Fluoro-3'-(trifluoromethyl)acetophenone
分子式: C₉H₆F₄O
分子量: 206.14 g/mol
MDL编号: MFCD00061252
CAS号: 208173-24-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83388e3b5e26a95f24a8fc705ca17f4d6891e6a2&vid=83388e3b5e26a95f24a8fc705ca17f4d6891e6a2&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR