2-Bromoethyl phenyl sulphide ≥97.0% (by GC)

供应商: TCI

Synonyms: (2-Bromoethyl)(phenyl)sulphane

B2085-25G B2085-5G
TCIAB2085-25GEA 0 CNY
TCIAB2085-25G TCIAB2085-5G
2-Bromoethyl phenyl sulphide ≥97.0% (by GC)
2-Bromoethyl phenyl sulphide
分子式: C₈H₉BrS
分子量: 217.12 g/mol
沸点: 137°C
存储条件: Refrigerator
MDL编号: MFCD00040826
CAS号: 4837-01-8
EINECS: 225-422-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02a981dca38aeabfc68f734487434c3b233bfbbe&vid=02a981dca38aeabfc68f734487434c3b233bfbbe&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR