2,4,6-Triisopropylphenylboronic acid (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: 2,4,6-Triisopropylbenzeneboronic acid, (2,4,6-Triisopropylphenyl)boronic acid

T2654-1G T2654-5G
TCIAT2654-1GEA 0 CNY
TCIAT2654-1G TCIAT2654-5G
2,4,6-Triisopropylphenylboronic acid (contains varying amounts of Anhydride)
2,4,6-Triisopropylphenylboronic acid
分子式: C₁₅H₂₅BO₂
MDL编号: MFCD02683099
CAS号: 154549-38-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba4eb5605a51dcbf4fa3e848190c1ca88650e209&vid=ba4eb5605a51dcbf4fa3e848190c1ca88650e209&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR