4-叔丁基苯硼酸 (contains varying amounts of Anhydride)
供应商: TCI
|
Warning
|
B2251-25G
B2251-5G
TCIAB2251-25GEA
0
CNY
TCIAB2251-25G
TCIAB2251-5G
4-叔丁基苯硼酸 (contains varying amounts of Anhydride)
4-叔丁基苯硼酸
分子式:
C₁₀H₁₅BO₂ |
MDL编号:
MFCD01009697 CAS号: 123324-71-0 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...