Sodium-1-tridecanesulphonate ≥98.0% (W)

供应商: TCI

T0756-25G T0756-5G
TCIAT0756-25GEA 0 CNY
TCIAT0756-25G TCIAT0756-5G
Sodium-1-tridecanesulphonate ≥98.0% (W)
Sodium-1-tridecanesulphonate
分子式: C₁₃H₂₇NaO₃S
MDL编号: MFCD00024993
CAS号: 5802-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fbbf5ba9c577a84d27b4e4c7918c003f8e6e069&vid=3fbbf5ba9c577a84d27b4e4c7918c003f8e6e069&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Gravimetric Method) min. 98.0 % (after drying)
Solubility in hot Water almost transparency
Drying loss max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR