Sodium-1-octadecanesulphonate

供应商: TCI

O0124-25G O0124-5G
TCIAO0124-25GEA 0 CNY
TCIAO0124-25G TCIAO0124-5G
Sodium-1-octadecanesulphonate
Sodium-1-octadecanesulphonate
分子式: C₁₈H₃₇NaO₃S
MDL编号: MFCD00051526
CAS号: 13893-34-0
EINECS: 237-660-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=819dfc50642673f13096a88d0001a447f802df95&vid=819dfc50642673f13096a88d0001a447f802df95&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Solubility in hot Water almost transparency
Drying loss max. 1.0 %
Ignition residue (Sulfate) 19.5 to 20.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR