1-壬烷磺酸钠 ≥98.0% (by titrimetric analysis) for ion pair chromatography
供应商: TCI
Synonyms:
1-壬烷磺酸
I0347-25G
I0347-5G
TCIAI0347-25GEA
0
CNY
TCIAI0347-25G
TCIAI0347-5G
1-壬烷磺酸钠 ≥98.0% (by titrimetric analysis) for ion pair chromatography
1-壬烷磺酸钠
分子式:
C₉H₁₉NaO₃S 分子量: 230.30 g/mol |
MDL编号:
MFCD00025010 CAS号: 35192-74-6 |
|
产品技术规格
Appearance | White powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Absorbance (210nm) | max. 1.000 |
Absorbance (230nm) | max. 0.300 |
Absorbance (254nm) | max. 0.130 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...