乙基乙烯基硫醚 ≥93.0% (by GC)

供应商: TCI
Danger

E0335-25ML E0335-5ML
TCIAE0335-25MLEA 0 CNY
TCIAE0335-25ML TCIAE0335-5ML
乙基乙烯基硫醚 ≥93.0% (by GC)
乙基乙烯基硫醚
分子式: C₄H₈S
沸点: 91°C
闪点: –6°C
MDL编号: MFCD00015180
CAS号: 627-50-9
UN号: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb60b1dfed5dc68d55ca30524e88ab3ec7547a25&vid=bb60b1dfed5dc68d55ca30524e88ab3ec7547a25&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR