1,3,5-Triacetylbenzene ≥98.0% (by GC)
供应商: TCI
Synonyms:
1,1',1''-(Benzene-1,3,5-triyl)triethanone
T1337-25G
T1337-5G
TCIAT1337-25GEA
0
CNY
TCIAT1337-25G
TCIAT1337-5G
1,3,5-Triacetylbenzene ≥98.0% (by GC)
1,3,5-Triacetylbenzene
分子式:
C₁₂H₁₂O₃ 分子量: 204.2200 g/mol |
MDL编号:
MFCD00008741 CAS号: 779-90-8 EINECS: 212-302-1 |
|
产品技术规格
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 162.0 to 165.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...