1,3-Diphenylacetone-p-toluenesulphonylhydrazone ≥98.0% (by HPLC, titration analysis)

供应商: TCI

D2093-10G D2093-25G
TCIAD2093-10GEA 0 CNY
TCIAD2093-10G TCIAD2093-25G
1,3-Diphenylacetone-p-toluenesulphonylhydrazone ≥98.0% (by HPLC, titration analysis)
1,3-Diphenylacetone-p-toluenesulphonylhydrazone
分子式: C₂₂H₂₂N₂O₂S
MDL编号: MFCD00009645
CAS号: 19816-88-7
EINECS: 243-344-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dfffd58f8101d4d1e268d838dce46a7179b8f42&vid=2dfffd58f8101d4d1e268d838dce46a7179b8f42&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR