4-Acetyldiphenyl sulphide ≥98.0% (通过GC)
供应商: TCI
|
Warning
|
A2355-25G
A2355-5G
TCIAA2355-25GEA
0
CNY
TCIAA2355-25G
TCIAA2355-5G
4-Acetyldiphenyl sulphide ≥98.0% (通过GC)
4-Acetyldiphenyl sulphide
分子式:
C₁₄H₁₂OS 沸点: 184°C |
MDL编号:
MFCD00026227 CAS号: 10169-55-8 EINECS: 233-443-5 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 65.0 to 69.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...