N-氟苯磺酰亚胺

供应商: LOCAL DEALS

Synonyms: N-氟代双苯磺酰胺, N-氟二苯磺酰胺, NFSI

R00831430124 R00831430121
CHLCR00831430124EA 0 CNY
CHLCR00831430124 CHLCR00831430121
N-氟苯磺酰亚胺
N-氟苯磺酰亚胺
分子式: C₁₂H₁₀FNO₄S₂
分子量: 315.34 g/mol
MDL编号: MFCD00144885
CAS号: 133745-75-2

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR