3-羧基苯硼酸 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 3-Boronobenzoic acid, 3-Carboxybenzeneboronic acid

344630010 344630050
ACRO344630010EA 0 CNY
ACRO344630010 ACRO344630050
3-羧基苯硼酸 97%
3-羧基苯硼酸
分子式: C₇H₇BO₄
溶点: 243…247 °C
存储条件: Ambient
MDL编号: MFCD00036833
CAS号: 25487-66-5

现在订购

产品技术规格

Appearance Off-white Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR