2-Carboxyphenylboronic acid (contains varying amounts of Anhydride)

供应商: TCI
Warning

C2501-1G C2501-5G
TCIAC2501-1GEA 0 CNY
TCIAC2501-1G TCIAC2501-5G
2-Carboxyphenylboronic acid (contains varying amounts of Anhydride)
分子式: C₇H₇BO₄
溶点: 162°C
存储条件: Refrigerator
MDL编号: MFCD01318118
CAS号: 149105-19-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dff2e09df3df02acc65721d23c94fb7b41906f0c&vid=dff2e09df3df02acc65721d23c94fb7b41906f0c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR