(R)-(+)-N-苄基-α-甲基苄胺 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: (R)-(+)-N-苄基-1-苯乙胺, (R)-(+)-N-(1-苯乙基)苄胺

B1528-25ML B1528-5ML
TCIAB1528-25MLEA 0 CNY
TCIAB1528-25ML TCIAB1528-5ML
(R)-(+)-N-苄基-α-甲基苄胺 ≥98.0% (by GC)
(R)-(+)-N-苄基-α-甲基苄胺
分子式: C₁₅H₁₇N
分子量: 211.31 g/mol
闪点: 158°C
MDL编号: MFCD00015010
CAS号: 38235-77-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=851fe6bc5fed0251e98423795d7b5a6b857cfcac&vid=851fe6bc5fed0251e98423795d7b5a6b857cfcac&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +38.0 to +42.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR