2-(三氟甲基)丙烯酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: TFMAA

T1512-25G T1512-5G
TCIAT1512-25GEA 0 CNY
TCIAT1512-25G TCIAT1512-5G
2-(三氟甲基)丙烯酸 ≥98.0% (by titrimetric analysis)
2-(三氟甲基)丙烯酸
分子式: C₄H₃F₃O₂
分子量: 140.06 g/mol
沸点: 148°C
MDL编号: MFCD00042424
CAS号: 381-98-6
UN号: 3261
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64a0ef59900d83bb3593d5c6f0469bf8dfc92fba&vid=64a0ef59900d83bb3593d5c6f0469bf8dfc92fba&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light red powder to crystaline
Purity (Neutralization titration) min. 98.0 %
Melting point 51.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR