Allyl ethyl carbonate ≥97.0% (通过GC)
供应商: TCI
|
Warning
|
C1280-25ML
C1280-5ML
TCIAC1280-25MLEA
0
CNY
TCIAC1280-25ML
TCIAC1280-5ML
Allyl ethyl carbonate ≥97.0% (通过GC)
Allyl ethyl carbonate
分子式:
C₆H₁₀O₃ 沸点: 149°C |
MDL编号:
MFCD00049113 CAS号: 1469-70-1 UN号: 3272 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...