Cholesterol nonyl carbonate

供应商: TCI

B0256-1G B0256-10G
TCIAB0256-1GEA 0 CNY
TCIAB0256-1G TCIAB0256-10G
Cholesterol nonyl carbonate
Cholesterol nonyl carbonate
分子式: C₃₇H₆₄O₃
MDL编号: MFCD00059506
CAS号: 15455-83-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8375f01215308c872a5ff2233184a14c6f0f5ab3&vid=8375f01215308c872a5ff2233184a14c6f0f5ab3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR