二糠基硫醚 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 二糠基硫化物

D3420-25G D3420-5G
TCIAD3420-25GEA 0 CNY
TCIAD3420-25G TCIAD3420-5G
二糠基硫醚 ≥98.0% (by GC)
二糠基硫醚
分子式: C₁₀H₁₀O₂S
分子量: 194.2500 g/mol
沸点: 143°C
MDL编号: MFCD00010081
CAS号: 13678-67-6
EINECS: 237-172-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5c5b0afbeaf808b174ce5d545939a590fd6f5cc&vid=c5c5b0afbeaf808b174ce5d545939a590fd6f5cc&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR