4'-Nitro-3'-(trifluoromethyl)acetanilide ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
N-(4-Nitro-3-(trifluoromethyl)phenyl)acetamide
N0548-25G
N0548-5G
TCIAN0548-25GEA
0
CNY
TCIAN0548-25G
TCIAN0548-5G
4'-Nitro-3'-(trifluoromethyl)acetanilide ≥98.0% (by GC)
4'-Nitro-3'-(trifluoromethyl)acetanilide
分子式:
C₉H₇F₃N₂O₃ 分子量: 248.1600 g/mol |
MDL编号:
MFCD00017994 CAS号: 393-12-4 |
|
产品技术规格
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 122.0 to 125.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...