Cut resistant gloves, ActivArmr® 80-600
供应商: Ansell
80-600/10 80-600/8 80-600/7 80-600/11 80-600/9
ANSE80-600/10EA 0 CNY
ANSE80-600/10 ANSE80-600/8 ANSE80-600/7 ANSE80-600/11 ANSE80-600/9
Cut resistant gloves, ActivArmr® 80-600
手套
These Kevlar® knitted gloves offer good cut resistance and robust overall mechanical performance.
  • Natural rubber coating on palm provides resistance to abrasion, tearing and punctures
  • Raised crinkle finish of the latex coating gives an excellent grip in both dry and wet conditions
  • Silicone-free - prevents fingerprints on metalised warm insulation glass, leaving surfaces clean and avoiding the need for extra cleaning work
Ideal for safe handling of glass and metal sheets, plastics and sharp edged objects.
Certifications: EN 388:2016 (3444C), EN 407 (X2XXXX); Cat. III.