聚酯密封膜
供应商: VWR
VWR®
VWRI731-0319EA 0 CNY
VWRI731-0319 VWRI731-0320
聚酯密封膜
密封件 微孔板封板膜和铝箔
聚酯膜,丙烯酸胶黏剂
该款49,5 µm厚的聚酯膜能够最大程度减少蒸发,避免孔内容物溢出和交叉污染。
  • Recommended temperature range from −40 to +120 °C
  • Available sterile and non sterile
  • Marginally pierceable by single-channel pipette
  • Applications: Short-term storage, incubation, ELISA
每个膜的长×深:141,0/79,4 mm