6,6'-Di-tert-butyl-4,4'-thiodi-m-cresol ≥98.0% (通过GC)

供应商: TCI
Warning

T0196-25G T0196-500G
TCIAT0196-25GEA 0 CNY
TCIAT0196-25G TCIAT0196-500G
6,6'-Di-tert-butyl-4,4'-thiodi-m-cresol ≥98.0% (通过GC)
6,6'-Di-tert-butyl-4,4'-thiodi-m-cresol
分子式: C₂₂H₃₀O₂S
MDL编号: MFCD00026287
CAS号: 96-69-5
EINECS: 202-525-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=511a9c1985a50ce01c09ff13720bbdeebfeec830&vid=511a9c1985a50ce01c09ff13720bbdeebfeec830&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 160.0 to 164.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR