4-(2-Thiazolylazo)resorcinol ≥99.0% (by titrimetric analysis) metal indicator, for spectrophotometry

供应商: TCI
Warning

A5002-1G A5002-5G
TCIAA5002-1GEA 0 CNY
TCIAA5002-1G TCIAA5002-5G
4-(2-Thiazolylazo)resorcinol ≥99.0% (by titrimetric analysis) metal indicator, for spectrophotometry
4-(2-Thiazolylazo)resorcinol
分子式: C₉H₇N₃O₂S
MDL编号: MFCD00005322
CAS号: 2246-46-0
EINECS: 218-836-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49c6fae784a1d49e189cd494d643df7e8e5c4dd3&vid=e2cd0682cd571cd973c82cbede4ed9c0102e8874&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in hot EtOH (50vol%) almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR