B-溴-1,2-苯二酚硼烷 ≥97.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
Synonyms:
B-溴邻苯二酚硼烷
B2132-25G
B2132-5G
TCIAB2132-25GEA
0
CNY
TCIAB2132-25G
TCIAB2132-5G
B-溴-1,2-苯二酚硼烷 ≥97.0% (by titrimetric analysis)
B-溴-1,2-苯二酚硼烷
分子式:
C₆H₄BBrO₂ 沸点: 76°C 溶点: 52°C 存储条件: Refrigerator |
MDL编号:
MFCD00013341 CAS号: 51901-85-0 UN号: 2925 ADR: 4.1 III |
|
产品技术规格
Appearance | White to Amber to Dark purple powder to lump |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...