(4-Nitrophenyl)-β-D-galactopyranoside 98,0 HPLC_ASSAY_METHOD Substrate for beta-Galactosidase
供应商: TCI
Synonyms:
pNPG, 4-Nitrophenyl β-D-Galactopyranoside
N0616-1G
N0616-5G
TCIAN0616-1GEA
0
CNY
TCIAN0616-1G
TCIAN0616-5G
(4-Nitrophenyl)-β-D-galactopyranoside 98,0 HPLC_ASSAY_METHOD Substrate for beta-Galactosidase
(4-Nitrophenyl)-β-D-galactopyranoside
分子式:
C₁₂H₁₅NO₈ 分子量: 301.25 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00063256 CAS号: 3150-24-1 EINECS: 221-584-5 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 179.0 to 183.0 deg-C |
Specific rotation [a]20/D | -80.0 to -87.0 deg (C=1, H2O) |
beta-Galactosidase detection test | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...