Ferbam (irontris(dimethyldithiocarbamate)) ≥97.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: Ferric dimethyldithiocarbamate, Ferbam, Iron(III) dimethyldithiocarbamate, Dimethyldithiocarbamic acid Ferric salt

D1267-25G D1267-500G
TCIAD1267-25GEA 0 CNY
TCIAD1267-25G TCIAD1267-500G
Ferbam (irontris(dimethyldithiocarbamate)) ≥97.0% (by titrimetric analysis)
Ferbam (irontris(dimethyldithiocarbamate))
分子式: C₉H₁₈FeN₃S₆
MDL编号: MFCD00067269
CAS号: 14484-64-1
EINECS: 238-484-2
UN号: 2811
ADR: 6.1 II
Merck Index: 14,04013
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4665205e495d535816276c2f0848c899abcdf7c7&vid=4665205e495d535816276c2f0848c899abcdf7c7&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Black powder to crystal
Purity (Chelometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR