1,4,10,13-四氧杂-7,16-二氮杂环十八烷 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 1,10-二氮杂-18-冠-6

D2323-1G D2323-5G
TCIAD2323-1GEA 0 CNY
TCIAD2323-1G TCIAD2323-5G
1,4,10,13-四氧杂-7,16-二氮杂环十八烷 ≥98.0% (by titrimetric analysis)
1,4,10,13-四氧杂-7,16-二氮杂环十八烷
分子式: C₁₂H₂₆N₂O₄
分子量: 262.35 g/mol
MDL编号: MFCD00005112
CAS号: 23978-55-4
EINECS: 245-965-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=413b7875125957b1ec236d1e257bb502ec775e3e&vid=413b7875125957b1ec236d1e257bb502ec775e3e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Dichloromethane very faint turbidity
Melting point 112.0 to 116.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR