4-甲氧基-3-硝基苯甲醛 ≥98.0% (通过GC)
供应商: TCI
|
Warning
|
N0830-25G
N0830-5G
TCIAN0830-25GEA
0
CNY
TCIAN0830-25G
TCIAN0830-5G
4-甲氧基-3-硝基苯甲醛 ≥98.0% (通过GC)
4-甲氧基-3-硝基苯甲醛
分子式:
C₈H₇NO₄ |
MDL编号:
MFCD00126498 CAS号: 31680-08-7 |
|
产品技术规格
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...