D-(-)-α-苯基甘氨酸 ≥99.0% (by titrimetric analysis)

供应商: TCI

Synonyms: (R)-(-)-2-苯基甘氨酸, R-(-)-α-氨基苯基乙酸, D-2-苯基甘氨酸

P0820-25G P0820-500G
TCIAP0820-25GEA 0 CNY
TCIAP0820-25G TCIAP0820-500G
D-(-)-α-苯基甘氨酸 ≥99.0% (by titrimetric analysis)
D-(-)-α-苯基甘氨酸
分子式: C₈H₉NO₂
分子量: 151.16 g/mol
MDL编号: MFCD00008061
CAS号: 875-74-1
EINECS: 212-876-3
Merck Index: 14,07291
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d03bc46f0be771a51452e99777468b4107817030&vid=d03bc46f0be771a51452e99777468b4107817030&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 99.0 %
Specific rotation [a]20/D -155 to -159 deg (C=1, 1mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR