2-氯-N,N-二甲基丙胺 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: β-(二甲基氨基)异丙基氯, 2-氯-1-(二甲基氨基)丙烷, N-(2-氯丙基)二甲胺, 2-(二甲基氨基)异丙基氯

D1665-25G D1665-500G
TCIAD1665-25GEA 0 CNY
TCIAD1665-25G TCIAD1665-500G
2-氯-N,N-二甲基丙胺 ≥98.0% (by titrimetric analysis)
2-氯-N,N-二甲基丙胺
分子式: C₅H₁₃Cl₂N
MDL编号: MFCD00012534
CAS号: 4584-49-0
EINECS: 224-971-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7feb1aa2ffd7d478685acbc1639e2ce7a137b9ee&vid=7feb1aa2ffd7d478685acbc1639e2ce7a137b9ee&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 187.0 to 190.0 deg-C
Solubility in Water within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR