5-甲基-2-噻吩基硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: 5-甲基-2-噻吩硼酸

M1850-1G M1850-5G
TCIAM1850-1GEA 0 CNY
TCIAM1850-1G TCIAM1850-5G
5-甲基-2-噻吩基硼酸 (contains varying amounts of Anhydride)
5-甲基-2-噻吩基硼酸
分子式: C₅H₇BO₂S
存储条件: Freezer
MDL编号: MFCD01318166
CAS号: 162607-20-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92c081da2a57c7ef1a5f6963c5d73fd52a423c01&vid=92c081da2a57c7ef1a5f6963c5d73fd52a423c01&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (Neutralization titration) 97.0 to 115.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR