Zinc(II) acetylacetonate ≥96.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: Bis(2,4-pentanedionato-O,O')zinc (II), Zinc acetylacetonate, Zinc (II) 2,4-pentanedionate, Zinc (II) bis(2,4-pentanedionato-O,O')

Z0002-25G Z0002-500G
TCIAZ0002-25GEA 0 CNY
TCIAZ0002-25G TCIAZ0002-500G
Zinc(II) acetylacetonate ≥96.0% (by titrimetric analysis)
Zinc(II) acetylacetonate
分子式: C₁₀H₁₄O₄Zn
分子量: 263.61 g/mol
MDL编号: MFCD00000035
CAS号: 14024-63-6
EINECS: 237-860-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=491465ede6dfa48e40d6c122067b3ef5c053813d&vid=491465ede6dfa48e40d6c122067b3ef5c053813d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 96.0 %
Solubility in hot Acetylacetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR