4,4,5,5,5-五氟-1-戊醇 ≥93.0% (by GC)

供应商: TCI
Warning

Synonyms: 4,4,5,5,5-Pentafluoropentanol

P1342-25G P1342-5G
TCIAP1342-25GEA 0 CNY
TCIAP1342-25G TCIAP1342-5G
4,4,5,5,5-五氟-1-戊醇 ≥93.0% (by GC)
4,4,5,5,5-五氟-1-戊醇
分子式: C₅H₇F₅O
沸点: 135°C
闪点: 63°C
MDL编号: MFCD00153224
CAS号: 148043-73-6
EINECS: 421-360-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acb54b31d93c662fabdd10e086ba84d833bd0431&vid=acb54b31d93c662fabdd10e086ba84d833bd0431&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR